Merck
  • Small molecule developmental screens reveal the logic and timing of vertebrate development.
Small molecule developmental screens reveal the logic and timing of vertebrate development.
Proceedings of the National Academy of Sciences of the United States of America (2000-11-23)
R T Peterson, B A Link, J E Dowling, S L Schreiber
ABSTRACT
Much has been learned about vertebrate development by random mutagenesis followed by phenotypic screening and by targeted gene disruption followed by phenotypic analysis in model organisms. Because the timing of many developmental events is critical, it would be useful to have temporal control over modulation of gene function, a luxury frequently not possible with genetic mutants. Here, we demonstrate that small molecules capable of conditional gene product modulation can be identified through developmental screens in zebrafish. We have identified several small molecules that specifically modulate various aspects of vertebrate ontogeny, including development of the central nervous system, the cardiovascular system, the neural crest, and the ear. Several of the small molecules identified allowed us to dissect the logic of melanocyte and otolith development and to identify critical periods for these events. Small molecules identified in this way offer potential to dissect further these and other developmental processes and to identify novel genes involved in vertebrate development.
Sigma-Aldrich
HDL Receptor SR-BI Inhibitor, BLT-1, The HDL Receptor SR-BI Inhibitor, BLT-1, also referenced under CAS 321673-30-7, controls the biological activity of HDL Receptor SR-B. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.