Merck
  • Glutathione S-transferases play a role in the detoxification of flumethrin and chlorpyrifos in Haemaphysalis longicornis.
Glutathione S-transferases play a role in the detoxification of flumethrin and chlorpyrifos in Haemaphysalis longicornis.
Parasites & vectors (2018-08-11)
Emmanuel Pacia Hernandez, Kodai Kusakisako, Melbourne Rio Talactac, Remil Linggatong Galay, Takeshi Hatta, Kozo Fujisaki, Naotoshi Tsuji, Tetsuya Tanaka
ABSTRACT
Haemaphysalis longicornis is a tick of importance to health, as it serves as a vector of several pathogens, including Theileria orientalis, Babesia ovata, Rickettsia japonica and the severe fever with thrombocytopenia syndrome virus (SFTSV). Presently, the major method of control for this tick is the use of chemical acaricides. The glutathione S-transferase (GST) system is one mechanism through which the tick metabolizes these acaricides. Two GSTs from H. longicornis (HlGST and HlGST2) have been previously identified. Enzyme kinetic studies were performed to determine the interaction of acaricides with recombinant H. longicornis GSTs. Recombinant HlGST activity was inhibited by flumethrin and cypermethrin, while recombinant HlGST2 activity was inhibited by chlorpyrifos and cypermethrin. Using real-time RT-PCR, the upregulation of the HlGST gene was observed upon exposure to sublethal doses of flumethrin, while the HlGST2 gene was upregulated when exposed to sublethal doses of chlorpyrifos. Sex and strain dependencies in the induction of GST gene expression by flumethrin were also observed. Knockdown of the HlGST gene resulted in the increased susceptibility of larvae and adult male ticks to sublethal doses of flumethrin and the susceptibility of larvae against sublethal doses of chlorpyrifos was increased upon knockdown of HlGST2. HlGST could be vital for the metabolism of flumethrin in larvae and adult male ticks, while HlGST2 is important in the detoxification of chlorpyrifos in larval ticks.
Product Description
Supelco
Coumaphos, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
Flumethrin, PESTANAL®, analytical standard
Pricing and availability is not currently available.