- Determination of piperidinium ionic liquid cations in environmental water samples by solid phase extraction and hydrophilic interaction liquid chromatography.
Determination of piperidinium ionic liquid cations in environmental water samples by solid phase extraction and hydrophilic interaction liquid chromatography.
This paper presents a novel analytical method for the determination of piperidinium ionic liquid cations in environmental water by hydrophilic interaction liquid chromatography and solid-phase extraction technology. The left standing, centrifuged and filtered river water samples were first purified and concentrated through the C18 solid phase extraction column, and eluted with 0.02mol/L hydrochloric acid prepared in methanol and deionized water (80/20, v/v). Then the eluents were analyzed by a hydrophilic column combined with 0.8mmol/L 1-propyl-3-methyl imidazolium tetrafluoroborate aqueous solution/acetonitrile (40/60, v/v) as the mobile phase and indirect ultraviolet detection. The detection limits of piperidinium cations were less than 0.4mg/L. The relative standard deviations were less than 0.6%. The method has been successfully applied to the determination of piperidinium cations in Songhua River water samples. Recoveries were 80.0%-98.3%. This research may provide a reference for studying the environmental effect of ionic liquids.