Merck
  • Zinc-bundle structure of the essential RNA polymerase subunit RPB10 from Methanobacterium thermoautotrophicum.
Zinc-bundle structure of the essential RNA polymerase subunit RPB10 from Methanobacterium thermoautotrophicum.
Proceedings of the National Academy of Sciences of the United States of America (2000-06-07)
C D Mackereth, C H Arrowsmith, A M Edwards, L P McIntosh
ABSTRACT
The RNA polymerase subunit RPB10 displays a high level of conservation across archaea and eukarya and is required for cell viability in yeast. Structure determination of this RNA polymerase subunit from Methanobacterium thermoautotrophicum reveals a topology, which we term a zinc-bundle, consisting of three alpha-helices stabilized by a zinc ion. The metal ion is bound within an atypical CX(2)CX(n)CC sequence motif and serves to bridge an N-terminal loop with helix 3. This represents an example of two adjacent zinc-binding Cys residues within an alpha-helix conformation. Conserved surface features of RPB10 include discrete regions of neutral, acidic, and basic residues, the latter being located around the zinc-binding site. One or more of these regions may contribute to the role of this subunit as a scaffold protein within the polymerase holoenzyme.
Sigma-Aldrich
RNA Polymerase II, RPB10 subunit, GST tagged human, recombinant, expressed in E. coli, ≥85% (SDS-PAGE)