Merck
  • Simultaneous identification of three pseudoallergic components in Danshen injection by using high-expression Mas-related G protein coupled receptor X2 cell membrane chromatography coupled online to HPLC-ESI-MS/MS.
Simultaneous identification of three pseudoallergic components in Danshen injection by using high-expression Mas-related G protein coupled receptor X2 cell membrane chromatography coupled online to HPLC-ESI-MS/MS.
Journal of separation science (2018-03-02)
Yuanyuan Lin, Cheng Wang, Yajing Hou, Wei Sun, Delu Che, Liu Yang, Tao Zhang, Meng Sun, Huaizhen He, Langchong He
ABSTRACT
Adverse drug reactions of Danshen injection mainly manifested as pseudoallergic reactions. In the present study, salvianolic acid A and a pair of geometric isomers (isosalvianolic acid C and salvianolic acid C) were identified as pseudoallergic components in Danshen injection by a high-expression Mas-related G protein coupled receptor X2 cell membrane chromatography coupled online with high-performance liquid chromatography with electrospray ionization tandem mass spectrometry. Their pseudoallergic activities were evaluated by in vitro assay, which were consistent with the retention times on the cell membrane chromatography column. Salvianolic acid C, the most outstanding compound, was further found to induce pseudoallergic reaction through Mas-related G protein coupled receptor X2. All the results above indicated that the system developed in this study is an effective method for simultaneously analyzing pseudoallergic components, even those with similar structures and the microcomponents in complex samples (salvianolic acid C in Danshen injection).
Sigma-Aldrich
MISSION® esiRNA, targeting human MRGPRX2
Pricing and availability is not currently available.
Sigma-Aldrich
Triton X-100, laboratory grade
Pricing and availability is not currently available.