Merck
  • Phase I/II trial of recombinant human granulocyte-macrophage colony-stimulating factor following allogeneic bone marrow transplantation.
Phase I/II trial of recombinant human granulocyte-macrophage colony-stimulating factor following allogeneic bone marrow transplantation.
Blood (1991-05-01)
J Nemunaitis, C D Buckner, F R Appelbaum, C S Higano, M Mori, J Bianco, C Epstein, J Lipani, J Hansen, R Storb
ABSTRACT
Forty-seven patients with hematologic neoplasia received recombinant human granulocyte-macrophage colony-stimulating factor (rhGM-CSF) by daily 2-hour infusion following allogeneic bone marrow transplantation from HLA-identical sibling donors in a phase I-II dose-escalation trial. Dose levels ranged from 30 to 500 micrograms/m2/d. At doses at or below 250 micrograms/m2/d, toxicity felt to be caused by rhGM-CSF was negligible. However, three of five patients treated with 500 micrograms/m2/d had unacceptable side effects caused by rhGM-CSF. Two different graft-versus-host disease (GVHD) prophylactic regimens were administered. Twenty-seven evaluable patients were administered regimens that did not contain methotrexate (MTX) (Group I) and reached an absolute neutrophil count of 1,000/microL by a median of day 14. In contrast, 18 patients who received GVHD prophylactic regimens containing MTX (Group II) reached an absolute neutrophil count of 1,000/microL on a median of day 20. Patients in Group I had fewer febrile days and, of those discharged, had shorter initial hospitalizations than patients in Group II. The overall incidence of severe acute GVHD (grade 2 or greater) in the rhGM-CSF-treated patients was 28% and was similar to that in historical "good risk" patients who did not receive rhGM-CSF. These preliminary data suggest rhGM-CSF is unlikely to exacerbate GVHD in HLA-identical sibling donor transplants and indicate the need for randomized trials of rhGM-CSF in allogeneic marrow transplant patients.
Sigma-Aldrich
GM-CSF from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
GM-CSF rat, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
Granulocyte-Macrophage Colony-Stimulating Factor human, GM-CSF, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture