Merck
  • Generation of an anti-angiogenic endothelial progenitor cell line via endostatin gene transfer.
Generation of an anti-angiogenic endothelial progenitor cell line via endostatin gene transfer.
Molecular medicine reports (2018-02-28)
Jing Ai, Jun-Hui Sun, Ting Wan, Jian Ma, Lei Feng, Ke Yao
ABSTRACT
The viability of endothelial progenitor cells (EPCs) as a therapeutic treatment for neovascularization (NV) was subject to investigation in the present study. Furthermore, endostatin has previously been demonstrated to be an inhibitor of angiogenesis and a suppressant of vascular leakage. The aim of the present study was to generate transgenic EPCs with anti‑angiogenic effects for the treatment of ocular NV. EPCs were obtained from rat peripheral blood samples and then verified. A lentiviral‑endostatin‑green fluorescent protein recombinant construct was generated and used to infect EPCs. Transfected cells were then subjected to puromycin selection. Reverse transcription‑quantitative polymerase chain reaction and a western blot assay were then applied in order to determine both the endostatin mRNA and protein expression levels, respectively. In addition, vascular endothelial growth factor (VEGF) expression levels were also detected in order to observe the anti‑angiogenic effect of the endostatin‑transfected EPCs. Following puromycin (1 µg/ml) selection for 4 days, a stable endostatin‑transfected EPC line was generated. In this stable endostatin-transfected EPC line, the expression levels of endostatin increased; whereas the expression levels of VEGF decreased. The results of the present study revealed that EPCs can be genetically modified to overexpress endostatin, which may provide the cells with an anti‑angiogenic effect via increased expression of endostatin and decreased expression of VEGF. Thus, EPCs genetically modified to overexpress endostatin may serve as a potential therapeutic agent for ocular NV treatment.
Sigma-Aldrich
Lectin from Ulex europaeus (gorse, furze), FITC conjugate, lyophilized powder