Merck
  • Acetyl-CoA promotes glioblastoma cell adhesion and migration through Ca2+-NFAT signaling.
Acetyl-CoA promotes glioblastoma cell adhesion and migration through Ca2+-NFAT signaling.
Genes & development (2018-04-21)
Joyce V Lee, Corbett T Berry, Karla Kim, Payel Sen, Taehyong Kim, Alessandro Carrer, Sophie Trefely, Steven Zhao, Sully Fernandez, Lauren E Barney, Alyssa D Schwartz, Shelly R Peyton, Nathaniel W Snyder, Shelley L Berger, Bruce D Freedman, Kathryn E Wellen
ABSTRACT
The metabolite acetyl-coenzyme A (acetyl-CoA) is the required acetyl donor for lysine acetylation and thereby links metabolism, signaling, and epigenetics. Nutrient availability alters acetyl-CoA levels in cancer cells, correlating with changes in global histone acetylation and gene expression. However, the specific molecular mechanisms through which acetyl-CoA production impacts gene expression and its functional roles in promoting malignant phenotypes are poorly understood. Here, using histone H3 Lys27 acetylation (H3K27ac) ChIP-seq (chromatin immunoprecipitation [ChIP] coupled with next-generation sequencing) with normalization to an exogenous reference genome (ChIP-Rx), we found that changes in acetyl-CoA abundance trigger site-specific regulation of H3K27ac, correlating with gene expression as opposed to uniformly modulating this mark at all genes. Genes involved in integrin signaling and cell adhesion were identified as acetyl-CoA-responsive in glioblastoma cells, and we demonstrate that ATP citrate lyase (ACLY)-dependent acetyl-CoA production promotes cell migration and adhesion to the extracellular matrix. Mechanistically, the transcription factor NFAT1 (nuclear factor of activated T cells 1) was found to mediate acetyl-CoA-dependent gene regulation and cell adhesion. This occurs through modulation of Ca2+ signals, triggering NFAT1 nuclear translocation when acetyl-CoA is abundant. The findings of this study thus establish that acetyl-CoA impacts H3K27ac at specific loci, correlating with gene expression, and that expression of cell adhesion genes are driven by acetyl-CoA in part through activation of Ca2+-NFAT signaling.
Sigma-Aldrich
MISSION® esiRNA, targeting human NFATC2