Merck
  • Comparison of methods of high-pressure freezing and automated freeze-substitution of suspension cells combined with LR White embedding.
Comparison of methods of high-pressure freezing and automated freeze-substitution of suspension cells combined with LR White embedding.
Histochemistry and cell biology (2010-11-11)
Margarita Sobol, Vlada V Philimonenko, Pavel Hozák
ABSTRACT
In this study we present an optimized method of high-pressure freezing and automated freeze-substitution of cultured human cells, followed by LR White embedding, for subsequent immunolabeling. Also, the influence of various conditions of the freeze-substitution procedures such as temperature, duration, and additives in the substitution medium on the preservation of cryo-immobilized cells was analyzed. The recommended approach combines (1) automated freeze-substitution for high reproducibility and minimizing human-derived errors; (2) minimal addition of contrasting and fixing agents; (3) easy-to-use LR White resin for embedment; (4) good preservation of nuclei and nucleoli which are usually the most difficult structures to effectively vitrify and saturate in a resin; and (5) preservation of antigens for sensitive immunogold labeling.
Sigma-Aldrich
LR white acrylic resins, Medium
Sigma-Aldrich
LR white resin, standard