Merck
  • PPAR-α agonists acutely inhibit Ca2+-independent PLA2 to reduce H2O2-induced contractions in aortae of spontaneously hypertensive rats.
PPAR-α agonists acutely inhibit Ca2+-independent PLA2 to reduce H2O2-induced contractions in aortae of spontaneously hypertensive rats.
American journal of physiology. Heart and circulatory physiology (2017-11-12)
Hui Chen, Ricky Y K Man, Susan W S Leung
ABSTRACT
Hypertension is associated with endothelial dysfunction, which favors the release of endothelium-derived contracting factors, including vasoconstrictor prostanoids and reactive oxygen species. Peroxisome proliferator-activated receptor-α (PPAR-α) agonists, clinically used as lipid-lowering drugs, possess antioxidant properties and exert beneficial effects in the vascular system. The present study aimed to identify the mechanism(s) underlying the acute effects of the PPAR-α agonists Wy14643 and fenofibate on endothelium-dependent contractions, in particular those related to oxidative stress, in the aorta of the spontaneously hypertensive rat (SHR). Aortic rings with and without endothelium of male SHRs and normotensive Wistar-Kyoto rats were suspended in organ chambers for isometric tension measurements and homogenized for enzyme activity assays. Contractions to acetylcholine in quiescent SHR aortae with endothelium were reduced by tiron (superoxide anion scavenger), diethyldithiocarbamic acid (superoxide dismutase inhibitor), and acute treatment with either Wy14643 or fenofibrate. Similarly to contractions evoked by acetylcholine, H2O2-induced increases in tension in SHR aortae involved, in succession, phospholipase A2 (PLA2), cyclooxygenase, and thromboxane-prostanoid receptors. Wy14643 or fenofibrate, by decreasing the activity of endothelial Ca2+-independent PLA2, attenuated the contractions to H2O2. In conclusion, the increased oxidative stress in the SHR aorta (mainly increased production of H2O2 and its partially reduced product, hydroxyl radical) contributed to acetylcholine-induced, endothelium-dependent contractions; PPAR-α agonists likely inhibit the H2O2-mediated contractions by inhibiting endothelial Ca2+-independent PLA2. The present study highlights the prospective therapeutic effects of PPAR-α agonists in improving endothelial function in hypertension and other vascular implications due to oxidative stress. NEW & NOTEWORTHY Peroxisome proliferator-activated receptor-α agonists, which are used clinically as lipid-lowering drugs, acutely reduce H2O2-induced contractions in aortae of hypertensive rats by inhibiting the activity of endothelial Ca2+-independent phospholipase A2. These vascular effects of peroxisome proliferator-activated receptor-α agonists suggest that they may help to prevent vascular complications under pathological conditions associated with oxidative stress.
Sigma-Aldrich
Valeryl Salicylate, ≥98% (HPLC)
Sigma-Aldrich
Arachidonyl trifluoromethyl ketone, oil, ≥97% (NMR)
Sigma-Aldrich
Bromoenol lactone, ≥98% (TLC)