Merck
Exposure to pesticides in ambient air.
Australian journal of public health (1995-08-01)
J Beard, V Westley-Wise, G Sullivan
ABSTRACT
Ambient air was monitored for pesticides at four sites in Coffs Harbour, a coastal town (population about 50,000) surrounded by banana plantations. Air was sampled continuously for five consecutive months during the peak agricultural spraying period using vacuum pumps set to sample one litre per minute through ORBO-42 absorption tubes. Six pesticides were detected: three organochlorines and three organophosphates. The most commonly detected pesticide (14 per cent of all samples) was chlorpyrifos (maximum detected level 208.0 ng/m3, mean 3.6 ng/m3). Heptachlor was detected in 7.1 per cent of all samples (maximum detected level 133 ng/m3, mean 2.7 ng/m3). Other pesticides were only rarely detected. The only pesticide applied by air in the district (propiconazole) was not detected. If international health guidelines are used as a yardstick, these levels of exposure appear unlikely to present an appreciable health risk. Chlorpyrifos detection was associated with low wind speed (P = 0.012) and high temperature (P = 0.015), and detection at one site was associated with detection at another (P < 0.001). Chlorpyrifos detection was also associated with domestic applications within the town area as reported by pesticide applicators (P = 0.045). Peak agricultural use of chlorpyrifos did not coincide with peak detection periods. None of the detected organochlorines is registered for agricultural use, although at the time, heptachlor was permitted for use as a domestic termiticide. Even in a semirural town with nearby widespread use of agricultural chemicals, community exposures to pesticides in ambient air may largely relate to their nonagricultural use.
Product Description
Supelco
ORBO 609 Amberlite XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
Supelco
ORBO 80 Coated Filter, with cassettes unassembled
Supelco
ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
Supelco
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 23 2-HMP on Amberlite XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea
Supelco
ORBO 65P (OVS) Amberlite XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea
Supelco
ORBO 24 2-HMP on Amberlite XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
Supelco
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
Supelco
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
ORBO 43 Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea