![Zoledronic acid monohydrate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/496/776/7c9ad506-21b0-4fd8-a3a4-95e4cff91fb0/640/7c9ad506-21b0-4fd8-a3a4-95e4cff91fb0.png)
Zoledronic acid monohydrate
![Zoledronic acid monohydrate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/496/776/7c9ad506-21b0-4fd8-a3a4-95e4cff91fb0/640/7c9ad506-21b0-4fd8-a3a4-95e4cff91fb0.png)
Synonym(s):
P,P′-[1-Hydroxy-2-(1H-imidazol-1-yl)ethylidene]bisphosphonic acid monohydrate, Zoledronate monohydrate, [1-hydroxy-2-(1H-imidazol-1-yl)ethane-1,1-diyl]bis(phosphonic acid) monohydrate
Empirical Formula (Hill Notation):
C5H10N2O7P2·H2O
CAS Number:
Molecular Weight:
290.10
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Quality Segment
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing