Skip to Content
Merck
Zinc di[bis(trifluoromethylsulfonyl)imide] 95%

Zinc di[bis(trifluoromethylsulfonyl)imide]

Zinc di[bis(trifluoromethylsulfonyl)imide] 95%
Synonym(s):
Zn(NTf2)2
Empirical Formula (Hill Notation):
C4F12N2O8S4Zn
CAS Number:
Molecular Weight:
625.68

Products

Shipping
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing