![Zidovudine for system suitability A CRS, European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/398/819/57941fb4-a75f-47a5-8ba8-fcef5bf0880e/640/57941fb4-a75f-47a5-8ba8-fcef5bf0880e.png)
Zidovudine for system suitability A
![Zidovudine for system suitability A CRS, European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/398/819/57941fb4-a75f-47a5-8ba8-fcef5bf0880e/640/57941fb4-a75f-47a5-8ba8-fcef5bf0880e.png)
Synonym(s):
3′-Azido-3′-deoxythymidine, AZT, Azidothymidine, ZDV, Zidovudine
Empirical Formula (Hill Notation):
C10H13N5O4
CAS Number:
Molecular Weight:
267.24
Products
Formula Weight
Manufacturer
Melting Point (°C)
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing