Skip to Content
Merck
Yu-Wasa Auxiliary 97%

Yu-Wasa Auxiliary

Yu-Wasa Auxiliary 97%
Synonym(s):
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline, 2,3,5,6,α,α,α-Heptafluoro-p-toluidine
Linear Formula:
CF3C6F4NH2
CAS Number:
Molecular Weight:
233.09

Products

Shipping
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing