![Vanillin isobutyrate ≥98%, FG](/deepweb/assets/sigmaaldrich/product/structures/811/188/9dadf535-2ae1-4402-afed-88c8620b86a6/640/9dadf535-2ae1-4402-afed-88c8620b86a6.png)
Vanillin isobutyrate
![Vanillin isobutyrate ≥98%, FG](/deepweb/assets/sigmaaldrich/product/structures/811/188/9dadf535-2ae1-4402-afed-88c8620b86a6/640/9dadf535-2ae1-4402-afed-88c8620b86a6.png)
Linear Formula:
(CH3)2CHCO2C6H3-2-(OCH3)-4-(CHO)
Products
Product Category
Brand
Biological Source
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing