Skip to Content
Merck
Uridine 5′-diphosphogalactose disodium salt ≥97.0%

Uridine 5′-diphosphogalactose disodium salt

Uridine 5′-diphosphogalactose disodium salt ≥97.0%
Synonym(s):
UDP-Gal, UDP-galactose
Empirical Formula (Hill Notation):
C15H22N2Na2O17P2
CAS Number:
Molecular Weight:
610.27

Products

Brand
Biological Source
Formula Weight
Physical Form
Purity
Tissue
Showing 1-1 of 1
Compare
Product Number
Description
Pricing