Skip to Content
Merck
Urethane analytical standard

Urethane

Urethane analytical standard
Synonym(s):
Carbamic acid ethyl ester, Ethyl carbamate, Ethylurethane
Linear Formula:
NH2COOC2H5
CAS Number:
Molecular Weight:
89.09
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing