Skip to Content
Merck
Tungstophosphoric acid hydrate cryst. EMPLURA®

Tungstophosphoric acid hydrate

Tungstophosphoric acid hydrate cryst. EMPLURA®
Synonym(s):
Tungstophosphoric acid hydrate
Linear Formula:
H3[P(W3O10)4] · x H2O
CAS Number:
Molecular Weight:
2880.05 (anhydrous basis)

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Showing 1-2 of 2