![TSKgel® SuperHM-L HPLC Column phase polystyrene-divinylbenzene, L × I.D. 15 cm × 6 mm, 3 μm particle size](/deepweb/assets/sigmaaldrich/product/images/342/651/db3a08d7-1f80-4c89-9dd4-e6aeb083f0b8/640/db3a08d7-1f80-4c89-9dd4-e6aeb083f0b8.jpg)
TSKgel® SuperHM-L (3 µm) HPLC Columns
![TSKgel® SuperHM-L HPLC Column phase polystyrene-divinylbenzene, L × I.D. 15 cm × 6 mm, 3 μm particle size](/deepweb/assets/sigmaaldrich/product/images/342/651/db3a08d7-1f80-4c89-9dd4-e6aeb083f0b8/640/db3a08d7-1f80-4c89-9dd4-e6aeb083f0b8.jpg)
Synonym(s):
TSKgel® SuperH Columns
Products
Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing