Skip to Content
Merck
Troxerutin certified reference material, pharmaceutical secondary standard

Troxerutin

Troxerutin certified reference material, pharmaceutical secondary standard
Synonym(s):
2-[3,4-bis(2-Hydroxyethoxy)phenyl]-3-[[6-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-5-hydroxy-7-(2-hydroxyethoxy)-4H-1-benzopyran-4-one, 7,3′,4′-tris[O-(2-hydroxyethyl)]rutin, Trihydroxyethylrutin, Troxerutinum, tri(hydroxyethyl)rutoside, trioxyethylrutin, vitaminP4, 3′,4′,7-Tris[O-(2-hydroxyethyl)]rutin, Troxerutin
Empirical Formula (Hill Notation):
C33H42O19
CAS Number:
Molecular Weight:
742.68

Products

Brand
Color
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade