![Triphenylbenzyloxymethylphosphonium chloride 97%](/deepweb/assets/sigmaaldrich/product/images/415/605/89587a0d-4282-41d7-99a9-e824036a5b84/640/89587a0d-4282-41d7-99a9-e824036a5b84.jpg)
Triphenylbenzyloxymethylphosphonium chloride
![Triphenylbenzyloxymethylphosphonium chloride 97%](/deepweb/assets/sigmaaldrich/product/images/415/605/89587a0d-4282-41d7-99a9-e824036a5b84/640/89587a0d-4282-41d7-99a9-e824036a5b84.jpg)
Synonym(s):
(Benzyloxymethyl)triphenylphosphonium chloride
Empirical Formula (Hill Notation):
C26H24ClOP
CAS Number:
Molecular Weight:
418.89
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reaction Type
Showing 1-1 of 1