Skip to Content
Merck
Trimethyl phosphite ≥97%

Trimethyl phosphite

Trimethyl phosphite ≥97%
Synonym(s):
Methyl phosphite, P(OMe)3, Trimethoxyphosphine, Trimethyl phosphite
Linear Formula:
(CH3O)3P
CAS Number:
Molecular Weight:
124.08

Products

Shipping
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Reaction Type
Reagent Type
Showing 1-3 of 3
Compare
Product Number
Description
Pricing