Skip to Content
Merck
Trimethoprim certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Trimethoprim

Trimethoprim certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine, NSC 106568
Empirical Formula (Hill Notation):
C14H18N4O3
CAS Number:
Molecular Weight:
290.32
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique