Trihexyphenidyl Related Compound A
Synonym(s):
1-Phenyl-3-(piperidin-1-yl)propan-1-one hydrochloride, 3-Piperidinopropiophenone hydrochloride
Empirical Formula (Hill Notation):
C14H19NO · HCl
CAS Number:
Molecular Weight:
253.77
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2