Skip to Content
Merck
Triflusal ≥98% (HPLC), powder

Triflusal

Triflusal ≥98% (HPLC), powder
Synonym(s):
Triflusal, α,α,α-trifluoro-2,4-cresotic acid acetate, 2-(Acetyloxy)-4-(trifluoromethyl)benzoic acid, Drisgen, acetyl-4-trifluoromethylsalicylic acid
Empirical Formula (Hill Notation):
C10H7F3O4
CAS Number:
Molecular Weight:
248.16

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing