
Triethylamine hydrochloride

Linear Formula:
(C2H5)3N · HCl
CAS Number:
Molecular Weight:
137.65
Products
Shipping
Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing