
Triamterene impurity B

Synonym(s):
2,7-Diamino-6-phenylpteridin-4-ol, 2,7-Diamino-4-hydroxy-6-phenylpteridine
Empirical Formula (Hill Notation):
C12H10N6O
CAS Number:
Molecular Weight:
254.25
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing