Skip to Content
Merck
Triacetin analytical standard

Triacetin

Triacetin analytical standard
Synonym(s):
1,2,3-Triacetoxypropane, 1,2,3-Triacetylglycerol, Glyceryl triacetate, Triacetin, Glycerol triacetate
Linear Formula:
(CH3COOCH2)2CHOCOCH3
CAS Number:
Molecular Weight:
218.20
EC Number:

Products

Shipping
Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique