Skip to Content
Merck
trans-Isoferulic acid analytical standard

trans-Isoferulic acid

trans-Isoferulic acid analytical standard
Synonym(s):
trans-3-(3-Hydroxy-4-methoxyphenyl)acrylic acid, trans-3-Hydroxy-4-methoxycinnamic acid, Hesperetic acid
Empirical Formula (Hill Notation):
C10H10O4
CAS Number:
Molecular Weight:
194.18
EC Number:

Products

Shipping
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing