Skip to Content
Merck
trans-2-Nonenal analytical standard

trans-2-Nonenal

trans-2-Nonenal analytical standard
Linear Formula:
CH3(CH2)5CH=CHCHO
CAS Number:
Molecular Weight:
140.22
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing