
trans-2-(4-Bromophenyl)vinylboronic acid MIDA ester

Empirical Formula (Hill Notation):
C13H13BBrNO4
Molecular Weight:
337.96
Products
Shipping
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing