Titanium(IV) oxysulfate
Synonym(s):
Titanyl sulfate
Linear Formula:
TiOSO4 · aq
CAS Number:
Molecular Weight:
159.93 (anhydrous basis)
Products
Shipping
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing