Skip to Content
Merck
Thioglycoll. B. clear 750ml ready-to-use, bottle volume 750 mL , filling volume

Thioglycoll. B. clear 750ml

Thioglycoll. B. clear 750ml ready-to-use, bottle volume 750 mL , filling volume
Synonym(s):
Fluid Thioglycollate Medium

Products

Product Category
Brand
Physical Form
Agency
Capacity (mL)
Volume (ml)
Quality Segment
Closure Type
Application
Microorganism Suitability
Package Size
Sterility
Sterilization Method
Showing 1-1 of 1