
Theacrine

Synonym(s):
1,3,7,9-Tetramethyluric acid, 1,3,7,9-Tetramethyl-2,6,8-trihydroxypurine
Empirical Formula (Hill Notation):
C9H12N4O3
CAS Number:
Molecular Weight:
224.22
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing