Skip to Content
Merck
Tetratriacontane analytical standard

Tetratriacontane

Tetratriacontane analytical standard
Linear Formula:
CH3(CH2)32CH3
CAS Number:
Molecular Weight:
478.92
EC Number:

Products

Shipping
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing