Skip to Content
Merck
Tetrabutylammonium fluoride trihydrate technical, ≥90% (T)

Tetrabutylammonium fluoride trihydrate

Tetrabutylammonium fluoride trihydrate technical, ≥90% (T)
Synonym(s):
TBAF
Linear Formula:
[CH3(CH2)3]4NF · 3H2O
CAS Number:
Molecular Weight:
315.51
EC Number:

Products

Shipping
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2