Skip to Content
Merck
(tert-Butyldimethylsilyl)acetylene 99%

(tert-Butyldimethylsilyl)acetylene

(tert-Butyldimethylsilyl)acetylene 99%
Synonym(s):
Ethynyl-tert-butyldimethylsilane
Linear Formula:
(CH3)3CSi(CH3)2C≡CH
CAS Number:
Molecular Weight:
140.30

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing