Skip to Content
Merck
Terephthalic acid analytical standard

Terephthalic acid

Terephthalic acid analytical standard
Synonym(s):
Benzene-1,4-dicarboxylic acid, Terephthalic acid
Linear Formula:
C6H4-1,4-(CO2H)2
CAS Number:
Molecular Weight:
166.13
EC Number:

Products

Shipping
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Markush Class
Markush Group
Monomer Type
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing