Skip to Content
Merck
Tenoxicam European Pharmacopoeia (EP) Reference Standard

Tenoxicam

Tenoxicam European Pharmacopoeia (EP) Reference Standard
Synonym(s):
4-Hydroxy-2-methyl-N-2-pyridinyl-2H-thieno(2,3-e)-1,2-thiazine-3-carboxamide 1,1-dioxide
Empirical Formula (Hill Notation):
C13H11N3O4S2
CAS Number:
Molecular Weight:
337.37

Products

Shipping
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3