![TAPS sodium salt ≥99% (titration)](/deepweb/assets/sigmaaldrich/product/structures/277/370/ce759c6f-8453-4095-ae7c-7fb895af69d4/640/ce759c6f-8453-4095-ae7c-7fb895af69d4.png)
TAPS sodium salt
![TAPS sodium salt ≥99% (titration)](/deepweb/assets/sigmaaldrich/product/structures/277/370/ce759c6f-8453-4095-ae7c-7fb895af69d4/640/ce759c6f-8453-4095-ae7c-7fb895af69d4.png)
Synonym(s):
N-tris(Hydroxymethyl)methyl-3-aminopropanesulfonic acid sodium-potassium salt, [(2-Hydroxy-1,1-bis(hydroxymethyl)ethyl)amino]-1-propanesulfonic acid sodium-potassium salt
Empirical Formula (Hill Notation):
C7H16NNaO6S
CAS Number:
Molecular Weight:
265.26
Products
Shipping
Product Category
Brand
Formula Weight
pH Value
Physical Form
pK Value
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing