Skip to Content
Merck
Taka-Diastase from Aspergillus oryzae powder, slightly beige, ~100 U/mg

Taka-Diastase from Aspergillus oryzae

Taka-Diastase from Aspergillus oryzae powder, slightly beige, ~100 U/mg
Synonym(s):
α-Amylase from Aspergillus oryzae, 1,4-α-D-Glucan-glucanohydrolase
EC Number:

Products

Product Category
Brand
Biological Source
Color
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing