![Tacrolimus monohydrate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/408/278/04252ce6-f5d5-4cd8-acf8-985e5954ef7c/640/04252ce6-f5d5-4cd8-acf8-985e5954ef7c.png)
Tacrolimus monohydrate
![Tacrolimus monohydrate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/408/278/04252ce6-f5d5-4cd8-acf8-985e5954ef7c/640/04252ce6-f5d5-4cd8-acf8-985e5954ef7c.png)
Synonym(s):
FK-506 monohydrate, Tacrolimus
Empirical Formula (Hill Notation):
C44H69NO12 · H2O
CAS Number:
Molecular Weight:
822.03
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing