Skip to Content
Merck
Sulfometuron methyl PESTANAL®, analytical standard

Sulfometuron methyl

Sulfometuron methyl PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C15H16N4O5S
CAS Number:
Molecular Weight:
364.38
EC Number:

Products

Shipping
Product Category
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing