Skip to Content
Merck
Sulfamethizole for system suitability CRS, European Pharmacopoeia (EP) Reference Standard

Sulfamethizole for system suitability

Sulfamethizole for system suitability CRS, European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Sulfamethizole, 4-Amino-N-(5-methyl-1,3,4-thiadiazol-2-yl)benzenesulfonamide, N1-(5-Methyl-1,3,4-thiadiazol-2-yl)sulfanilamide, Sulfamethylthiadiazole
Empirical Formula (Hill Notation):
C9H10N4O2S2
CAS Number:
Molecular Weight:
270.33

Products

Formula Weight
Manufacturer
Quality Segment
API Family
Reference Material Type
Showing 1-1 of 1