Skip to Content
Merck
Sulconazole Nitrate certified reference material, pharmaceutical secondary standard

Sulconazole nitrate

Sulconazole Nitrate certified reference material, pharmaceutical secondary standard
Synonym(s):
Sulconazole nitrate salt, 1-(2-[p-Chlorobenzylthio]-2-[2,4-dichlorophenyl]ethyl)-1H-imidazole
Empirical Formula (Hill Notation):
C18H15Cl3N2S · HNO3
CAS Number:
Molecular Weight:
460.76

Products

Shipping
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade