Streptomycin sulfate for identification
Synonym(s):
Streptomycin sulfate salt
Linear Formula:
C21H39N7O12 · 1.5 H2SO4
CAS Number:
Molecular Weight:
728.69
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing