Sodium bicarbonate-13C
Synonym(s):
Sodium hydrogencarbonate-13C
Linear Formula:
NaH13CO3
CAS Number:
Molecular Weight:
85.00
Products
Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Special Grade
Showing 1-3 of 3