Skip to Content
Merck
Sodium acetate-2-13C,d3 99 atom % 13C, 50-60 atom % D

Sodium acetate-2-13C,d3

Sodium acetate-2-13C,d3 99 atom % 13C, 50-60 atom % D
Synonym(s):
Acetic acid-2-13C,2,2,2-d3 sodium salt
Linear Formula:
13CD3CO2Na
CAS Number:
Molecular Weight:
86.04

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-2 of 2