Skip to Content
Merck
Silicon phthalocyanine dichloride Dye content ~85 %

Silicon phthalocyanine dichloride

Silicon phthalocyanine dichloride Dye content ~85 %
Synonym(s):
Dichloro(29H,31H-phthalocyaninato)silicon, Phthalocyaninato-dichlorosilane
Empirical Formula (Hill Notation):
C32H16Cl2N8Si
CAS Number:
Molecular Weight:
611.51
EC Number:

Products

Shipping
Product Category
Formula Weight
Physical Form
Chemical Composition
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing